Learn More
Medchemexpress LLC Oct3/4-inducer-1 | 1016535-83-3 | 98.1% | 241.26 | 5 MG
SDP
Catalog No. 5000388011
Encompass_Preferred
Oct3/4-inducer-1 is a potent Oct3/4 inducer. It promotes expression and stabilization of Oct3/4, and enhances its transcriptional activity in diverse human somatic cells.
  • Promotes expression of Oct3/4.
  • Stabilizes Oct3/4.
  • Enhances transcriptional activity in diverse human somatic cells.
  • Stable under recommended storage conditions.
  • Suitable for research use only.
Catalog No. 50-003-88011 Supplier Medchemexpress LLC Supplier No. HY187735MG
Oct3/4-inducer-1 is a potent Oct3/4 inducer. It promotes expression and stabilization of Oct3/4, and enhances its transcriptional activity in diverse human somatic cells.
  • Promotes expression of Oct3/4.
  • Stabilizes Oct3/4.
  • Enhances transcriptional activity in diverse human somatic cells.
  • Stable under recommended storage conditions.
  • Suitable for research use only.