Learn More
Medchemexpress LLC Oct3/4-inducer-1 | 1016535-83-3 | 98.12% | 241.26 | 50 MG
SDP
Catalog No. 5000387965
Encompass_Preferred
Oct3/4-inducer-1 (compound 2) is a potent Oct3/4 inducer. It promotes the expression and stabilization of Oct3/4 and enhances its transcriptional activity in diverse human somatic cells.
  • Potent Oct3/4 inducer.
  • Promotes expression and stabilization of Oct3/4.
  • Enhances transcriptional activity in diverse human somatic cells.
  • For research use only.
Catalog No. 50-003-87965 Supplier Medchemexpress LLC Supplier No. HY1877350MG
Oct3/4-inducer-1 (compound 2) is a potent Oct3/4 inducer. It promotes the expression and stabilization of Oct3/4 and enhances its transcriptional activity in diverse human somatic cells.
  • Potent Oct3/4 inducer.
  • Promotes expression and stabilization of Oct3/4.
  • Enhances transcriptional activity in diverse human somatic cells.
  • For research use only.