Learn More
Medchemexpress LLC Octaarginine acetate | 98.3% | 1267.50 | 5 MG
SDP
Catalog No. 5000424199
Encompass_Preferred
Octaarginine acetate is a cell-penetrating peptide and proteasome inhibitor. It exhibits mixed-type inhibition against 20S proteasome chymotrypsin-like, caspase-like, and trypsin-like activities, and inhibits 26S proteasome activity with decreased efficiency. It induces ubiquitin-conjugated protein accumulation, mediates HSPG-dependent cellular internalization via macropinocytosis, and enhances liposomal cargo uptake and gene delivery. Octaarginine acetate can be used for the research of cervix carcinoma, collagen antibody-induced arthritis, and bacterial infections.
  • Potently inhibits chymotrypsin-like and caspase-like activities of 20S proteasomes.
  • Efficiently delivers protein, peptide, and plasmid DNA cargos into cultured cells.
  • Inhibits the growth of E. coli and S. aureus bacterial cells.
  • Inhibits HeLa human cancer cell growth in a concentration-dependent manner.
Catalog No. 50-004-24199 Supplier Medchemexpress LLC Supplier No. HYP2483B5MG
Add to Cart
Add to Cart
Octaarginine acetate is a cell-penetrating peptide and proteasome inhibitor. It exhibits mixed-type inhibition against 20S proteasome chymotrypsin-like, caspase-like, and trypsin-like activities, and inhibits 26S proteasome activity with decreased efficiency. It induces ubiquitin-conjugated protein accumulation, mediates HSPG-dependent cellular internalization via macropinocytosis, and enhances liposomal cargo uptake and gene delivery. Octaarginine acetate can be used for the research of cervix carcinoma, collagen antibody-induced arthritis, and bacterial infections.
  • Potently inhibits chymotrypsin-like and caspase-like activities of 20S proteasomes.
  • Efficiently delivers protein, peptide, and plasmid DNA cargos into cultured cells.
  • Inhibits the growth of E. coli and S. aureus bacterial cells.
  • Inhibits HeLa human cancer cell growth in a concentration-dependent manner.