Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Octachlorocyclopentene 98.0+%, TCI America™
SDP

Catalog No. O01095G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O01095G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O01095G Supplier TCI America Supplier No. O01095G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 706-78-5
Molecular Formula C5Cl8
Molecular Weight (g/mol) 343.655
MDL Number MFCD00145025
InChI Key DMZRCHJVWAKCAX-UHFFFAOYSA-N
Synonym Perchlorocyclopentene
PubChem CID 12814
IUPAC Name 1,2,3,3,4,4,5,5-octachlorocyclopentene
SMILES C1(=C(C(C(C1(Cl)Cl)(Cl)Cl)(Cl)Cl)Cl)Cl

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 343.66
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Octachlorocyclopentene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.