Learn More
Medchemexpress LLC Octacosane-d58 | 16416-33-4 | MFCD00190493 | 99.1% | 453.12 g/mol | C28D58 | 10 MG
SDP
Catalog No. 5000281984
Encompass_Preferred
Octacosane-d58 is the deuterium-labeled analogue of octacosane provided as a white to off-white solid for analytical and research applications. It is intended as a stable-isotope internal standard and tracer for mass spectrometry and chromatography, enabling reliable quantitation and method development in lipidomics and related workflows.
  • High isotopic labeling and overall purity: 99.1%.
  • Molecular formula C28D58 and molecular weight 453.12 g/mol.
  • Solid, white to off-white appearance for easy handling and storage.
  • Suitable as a mass spectrometry internal standard and tracer in quantitative analyses.
  • Available in small, analytical-scale package sizes for method development.
Catalog No. 50-002-81984 Supplier Medchemexpress LLC Supplier No. HYW272217S10MG
Octacosane-d58 is the deuterium-labeled analogue of octacosane provided as a white to off-white solid for analytical and research applications. It is intended as a stable-isotope internal standard and tracer for mass spectrometry and chromatography, enabling reliable quantitation and method development in lipidomics and related workflows.
  • High isotopic labeling and overall purity: 99.1%.
  • Molecular formula C28D58 and molecular weight 453.12 g/mol.
  • Solid, white to off-white appearance for easy handling and storage.
  • Suitable as a mass spectrometry internal standard and tracer in quantitative analyses.
  • Available in small, analytical-scale package sizes for method development.