Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Octacosane-d58 | 16416-33-4 | MFCD00190493 | 99.1% | 453.12 g·mol⁻¹ | C28D58 | 100 MG
SDP

Catalog No. 5000282016
Encompass_Preferred

Octacosane-d58 is the deuterium-labeled form of octacosane provided as a stable isotope analytical standard for research and analytical applications. It is commonly used as an internal standard in mass spectrometry and for isotope-tracing studies, retaining the chemical properties of octacosane while containing deuterium atoms.

  • Deuterium-labeled octacosane for isotope-labeling and tracing studies.
  • Intended as an analytical internal standard for mass spectrometry.
  • High purity: 99.05% (reported).
  • Chemical formula C28D58; molecular weight 453.12 g·mol⁻¹.
  • Available as a 100 mg solid package.
  • CAS number 16416-33-4 for unambiguous identification.

Catalog No. 50-002-82016 Supplier Medchemexpress LLC Supplier No. HYW272217S100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Octacosane-d58 is the deuterium-labeled form of octacosane provided as a stable isotope analytical standard for research and analytical applications. It is commonly used as an internal standard in mass spectrometry and for isotope-tracing studies, retaining the chemical properties of octacosane while containing deuterium atoms.

  • Deuterium-labeled octacosane for isotope-labeling and tracing studies.
  • Intended as an analytical internal standard for mass spectrometry.
  • High purity: 99.05% (reported).
  • Chemical formula C28D58; molecular weight 453.12 g·mol⁻¹.
  • Available as a 100 mg solid package.
  • CAS number 16416-33-4 for unambiguous identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.