Learn More
Medchemexpress LLC Octacosane-d58 | 16416-33-4 | MFCD00190493 | 99.1% | 453.12 g/mol | C28D58 | 25 MG
SDP
Supplier:  Medchemexpress LLC HYW272217S25MG
Encompass_Preferred
Octacosane-d58 is the deuterium-labeled analogue of octacosane used as a stable isotope standard for analytical and research applications. It is a high-purity, fully deuterated straight-chain hydrocarbon (C28D58, MW 453.12) suitable as an internal standard or tracer in mass spectrometry and metabolic studies. CAS: 16416-33-4.
  • High isotopic enrichment suitable for quantitative mass spectrometry.
  • High chemical purity for analytical consistency.
  • Fully deuterated C28 hydrocarbon provides a clear mass shift from unlabeled analogue.
  • Solid, stable material with extended shelf life when stored properly.
  • Supplied in small, weighed quantities for trace-level analyses.
Catalog No. 50-002-82240