Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Octacosanoic Acid 99+%, Thermo Scientific™

Catalog No. AC258200010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC258200010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC258200010 Supplier Thermo Scientific Chemicals Supplier No. 258200010

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 506-48-9
Molecular Formula C28H56O2
Molecular Weight (g/mol) 424.754
InChI Key UTOPWMOLSKOLTQ-UHFFFAOYSA-N
Synonym montanic acid, n-octacosanoic acid, octacosoic acid, unii-4bkl1a0kjy, 4bkl1a0kjy, n-octacosanoate, octacosoate, octacosansaeure, octacosanoicacid, octaeicosanoic acid
PubChem CID 10470
ChEBI CHEBI:31001
IUPAC Name octacosanoic acid
SMILES CCCCCCCCCCCCCCCCCCCCCCCCCCCC(=O)O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 91°C to 94°C
Color White
Density 0.81
Infrared Spectrum Authentic
Linear Formula CH3(CH2)26COOH
Quantity 1 g
Formula Weight 424.76
Percent Purity ≥99.0%
Physical Form Fine Crystalline Powder
Chemical Name or Material Octacosanoic acid, 99+%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.