Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Octacosanoic Acid 98.0+%, TCI America™
SDP

Catalog No. M1342100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1342100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1342100MG Supplier TCI America Supplier No. M1342100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 506-48-9
Molecular Formula C28H56O2
Molecular Weight (g/mol) 424.754
MDL Number MFCD00002812
InChI Key UTOPWMOLSKOLTQ-UHFFFAOYSA-N
Synonym montanic acid, n-octacosanoic acid, octacosoic acid, unii-4bkl1a0kjy, 4bkl1a0kjy, n-octacosanoate, octacosoate, octacosansaeure, octacosanoicacid, octaeicosanoic acid
PubChem CID 10470
ChEBI CHEBI:31001
IUPAC Name octacosanoic acid
SMILES CCCCCCCCCCCCCCCCCCCCCCCCCCCC(=O)O

Specifications

Melting Point 89°C
Color White
Quantity 100 mg
Formula Weight 424.75
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Octacosanoic Acid
EINECSNumber (2)-0419&(2)-0608
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.