Learn More
Cayman Chemical Octacosanoic Acid methyl ester, Purity: ≥95%, CAS: 55682-92-3, 50 mg
SDP
Catalog No. 502935366
Encompass_Preferred
A very long-chain fatty acid methyl ester; has been found in transesterified palm oil, biodiesel produced by the microalga Botryococcus, and in sediment samples from Lake Kivu in the East African rift valley
Catalog No. 50-293-5366 Supplier Cayman Chemical Supplier No. 2672650 MG
A very long-chain fatty acid methyl ester; has been found in transesterified palm oil, biodiesel produced by the microalga Botryococcus, and in sediment samples from Lake Kivu in the East African rift valley