Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Octadecafluorodecahydronaphthalene

Catalog No. 7726450ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
50 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
77-264-50ML 50 mL
77-264-250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 77-264-50ML Supplier Sigma Aldrich Supplier No. 7726450ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Octadecafluorodecahydronaphthalene

Specifications

CAS 306-94-5
Density 1.941 g/mL at 20°C (literature); 1.941 g/mL (at 20°C)
Boiling Point 141°C to 143°C (lit.)
Molecular Formula C10F18
Refractive Index n20/D 1.314 (literature)
Linear Formula C10F18
MDL Number MFCD00010626
Quantity 50 mL
Synonym Perfluorodecahydronaphthalene; Perfluorodecalin
Molecular Weight (g/mol) 462.08
Percent Purity ≥94% (GC)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.