Learn More
Medchemexpress LLC HY-W004307 100mg Medchemexpress, Octadecanal CAS:638-66-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW004307100MG
Encompass_Preferred
Medchemexpress, HY-W004307 100mg Octadecanal CAS:638-66-4 Octadecanal is a long-chain aldehyde, present in both thigh and breast muscle. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3221