Learn More
Medchemexpress LLC Octadecanal | 638-66-4 | 268.48 | 5 G
SDP
Catalog No. 5000447672
Encompass_Preferred
Octadecanal (Stearyl aldehyde) is a long-chain aldehyde component found in the phospholipid fractions of thigh and breast muscles of broiler chickens.
  • White to off-white solid
  • Store at -20°C under nitrogen
  • In solvent, store at -80°C for 6 months or -20°C for 1 month under nitrogen
  • Classified as ketones, aldehydes, and acids
  • Derived from endogenous metabolite
Catalog No. 50-004-47672 Supplier Medchemexpress LLC Supplier No. HYW0043075G
Octadecanal (Stearyl aldehyde) is a long-chain aldehyde component found in the phospholipid fractions of thigh and breast muscles of broiler chickens.
  • White to off-white solid
  • Store at -20°C under nitrogen
  • In solvent, store at -80°C for 6 months or -20°C for 1 month under nitrogen
  • Classified as ketones, aldehydes, and acids
  • Derived from endogenous metabolite