Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Octadecanal 95.0+%, TCI America™
SDP

Catalog No. O03681G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O03681G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O03681G Supplier TCI America Supplier No. O03681G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 638-66-4
Molecular Formula C18H36O
Molecular Weight (g/mol) 268.485
InChI Key FWWQKRXKHIRPJY-UHFFFAOYSA-N
Synonym Octadecanaldehyde, Octadecyl Aldehyde, Stearaldehyde, Stearyl Aldehyde
PubChem CID 12533
ChEBI CHEBI:17034
IUPAC Name octadecanal
SMILES CCCCCCCCCCCCCCCCCC=O

Specifications

Melting Point 46°C
Color White
Boiling Point 215°C
Quantity 1 g
Formula Weight 268.49
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Octadecanal
EINECSNumber (2)-0494
RTECSNumber RG0181500
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.