Learn More
Medchemexpress LLC Octadecane-1-thiol-d37 | 668433-57-6 | 98.1% | 323.79 | 1 MG
SDP
Catalog No. 5000451669
Encompass_Preferred
Octadecane-1-thiol-d37 is the deuterium labeled Octadecane-1-thiol. It is for research use only and not sold to patients.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process.
  • Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-51669 Supplier Medchemexpress LLC Supplier No. HYW009299S1MG
Octadecane-1-thiol-d37 is the deuterium labeled Octadecane-1-thiol. It is for research use only and not sold to patients.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process.
  • Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.