Learn More
Medchemexpress LLC Octadecane-1-thiol-d37 | 668433-57-6 | 98.1% | 323.79 | 5 MG
SDP
Catalog No. 5000366959
Encompass_Preferred
Octadecane-1-thiol-d37 is the deuterium labeled Octadecane-1-thiol. It is for research use only and not sold to patients. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Isotope-labeled compound: deuterium (2H, D)
Catalog No. 50-003-66959 Supplier Medchemexpress LLC Supplier No. HYW009299S5MG
Octadecane-1-thiol-d37 is the deuterium labeled Octadecane-1-thiol. It is for research use only and not sold to patients. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Isotope-labeled compound: deuterium (2H, D)