Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Octadecane

Catalog No. 502891475
Encompass
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-289-1475 500 g
O652100G 100 g
O65225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 50-289-1475 Supplier Sigma Aldrich Supplier No. O652500G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Octadecane
TRUSTED_SUSTAINABILITY

Specifications

CAS 593-45-3
Melting Point 26°C to 29°C (lit.)
Density 0.777 g/mL (at 25°C (literature))
Boiling Point 317°C (lit.)
Molecular Formula C18H38
Linear Formula CH3(CH2)16 CH3
MDL Number MFCD00009007
Quantity 500 g
Molecular Weight (g/mol) 254.49
Percent Purity 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.