Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Octadecane

Catalog No. O65225G
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O65225G 25 g
50-289-1475 500 g
O652100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. O65225G Supplier Sigma Aldrich Supplier No. O65225G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Octadecane

Specifications

CAS 593-45-3
Melting Point 26°C to 29°C (lit.)
Density 0.777 g/mL (at 25°C (literature))
Boiling Point 317°C (lit.)
Molecular Formula C18H38
Linear Formula CH3(CH2)16 CH3
MDL Number MFCD00009007
Quantity 25 g
Molecular Weight (g/mol) 254.49
Percent Purity 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.