Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Organic Chemistry Octadecane | 500G | 593-45-3 | MFCD00009007

Catalog No. O652500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred

Octadecane , 500G

About this item:

CAS #: 593-45-3
MDL #: MFCD00009007
Purity: 99%
Chemical Formula: C18H38
Molecular Weight: 254.49
UNSPSC Code: 12352100

Catalog No. O652500G Supplier Sigma Organic Chemistry Supplier No. O652500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Octadecane , 500G

About this item:

CAS #: 593-45-3
MDL #: MFCD00009007
Purity: 99%
Chemical Formula: C18H38
Molecular Weight: 254.49
UNSPSC Code: 12352100

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.