Learn More
Medchemexpress LLC Octadecanoic acid, 10-hydroxy- | 638-26-6 | MFCD08693364 | 99.8% | 300.48 g/mol | C18H36O3 | 25 MG
SDP
Catalog No. 5000282624
Encompass_Preferred
10-Hydroxystearic acid (10-HSA) is a hydroxylated stearic acid provided as a research reagent. It functions as a PPARα agonist and has been reported to stimulate collagen synthesis and exhibit anti-aging effects such as reducing the appearance of pores and hyperpigmentation. Supplied at high purity for in vitro and formulation studies.
Catalog No. 50-002-82624 Supplier Medchemexpress LLC Supplier No. HYW12815625MG
10-Hydroxystearic acid (10-HSA) is a hydroxylated stearic acid provided as a research reagent. It functions as a PPARα agonist and has been reported to stimulate collagen synthesis and exhibit anti-aging effects such as reducing the appearance of pores and hyperpigmentation. Supplied at high purity for in vitro and formulation studies.