Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Octadecanoic-d35 acid, 1,2,3-propanetriyl-d5 ester | 33048-69-0 | 99.1% | 10MG
SDP

Catalog No. 5000342171
Encompass_Preferred

Tristearin-d40 (Octadecanoic-d35 acid, 1,2,3-propanetriyl-d5 ester) is a stable isotope chemical identified by CAS number 33048-69-0. This compound presents as a white to off-white solid with a high purity of 99.12% as determined by HPLC. Its molecular formula is C57D110O6, and it has a molecular weight of 1002.16.

  • High purity of 99.12% (HPLC)
  • White to off-white solid appearance
  • Stable isotope compound for research applications
  • Molecular formula C57D110O6
  • Molecular weight 1002.16
  • Optimal powder storage: -20°C for 3 years; 4°C for 2 years
  • Optimal in solvent storage: -80°C for 6 months; -20°C for 1 month

Catalog No. 50-003-42171 Supplier Medchemexpress LLC Supplier No. HY127035S210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tristearin-d40 (Octadecanoic-d35 acid, 1,2,3-propanetriyl-d5 ester) is a stable isotope chemical identified by CAS number 33048-69-0. This compound presents as a white to off-white solid with a high purity of 99.12% as determined by HPLC. Its molecular formula is C57D110O6, and it has a molecular weight of 1002.16.

  • High purity of 99.12% (HPLC)
  • White to off-white solid appearance
  • Stable isotope compound for research applications
  • Molecular formula C57D110O6
  • Molecular weight 1002.16
  • Optimal powder storage: -20°C for 3 years; 4°C for 2 years
  • Optimal in solvent storage: -80°C for 6 months; -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.