Learn More
Medchemexpress LLC Octadecanoic-d35 acid, 1,2,3-propanetriyl-d5 ester | 33048-69-0 | 99.1% | 25 MG
SDP
Catalog No. 5000341874
Encompass_Preferred
Tristearin-d40 is a stable isotope, also known as Octadecanoic-d35 acid, 1,2,3-propanetriyl-d5 ester. This product is a white to off-white solid, demonstrating high purity and structural consistency.
  • White to off-white solid appearance
  • Consistent 1H NMR spectrum
  • Purity of 99.12% by HPLC
  • Stable as a powder at -20°C for 3 years or 4°C for 2 years
  • Stable in solvent at -80°C for 6 months or -20°C for 1 month
Catalog No. 50-003-41874 Supplier Medchemexpress LLC Supplier No. 25MGHY127035S2
Tristearin-d40 is a stable isotope, also known as Octadecanoic-d35 acid, 1,2,3-propanetriyl-d5 ester. This product is a white to off-white solid, demonstrating high purity and structural consistency.
  • White to off-white solid appearance
  • Consistent 1H NMR spectrum
  • Purity of 99.12% by HPLC
  • Stable as a powder at -20°C for 3 years or 4°C for 2 years
  • Stable in solvent at -80°C for 6 months or -20°C for 1 month