Learn More
Apexbio Technology LLC Octadecanoyl-carnitine-d3-HCl
SDP
Catalog No. 5000570421
Encompass_Preferred
Octadecanoyl carnitine-d3 HCl is a deuterated fatty acid ester involved in biomedical research primarily focused on lipid metabolism and transport mechanisms Originating from studies of acylcarnitines it is instrumental in examining fatty acid oxidation pathways and mitochondrial function This compound targets carnitine acyltransferases playing a crucial role in translocating acyl groups across cellular membranes Its activity is commonly assessed in the nanomolar to low micromolar range This makes it a valuable tool for in vitro assays involving various cell models including those related to metabolic disorders Additionally it is utilized in animal studies to explore its potential effects on energy metabolism and lipid-related conditions Drug screening efforts employ it to evaluate new therapeutic candidates targeting metabolic pathways Experimental concentrations typically depend on the specific research design employed ensuring relevance and accuracy in findings
Catalog No. 50-005-70421 Supplier Apexbio Technology LLC Supplier No. SL10261
Octadecanoyl carnitine-d3 HCl is a deuterated fatty acid ester involved in biomedical research primarily focused on lipid metabolism and transport mechanisms Originating from studies of acylcarnitines it is instrumental in examining fatty acid oxidation pathways and mitochondrial function This compound targets carnitine acyltransferases playing a crucial role in translocating acyl groups across cellular membranes Its activity is commonly assessed in the nanomolar to low micromolar range This makes it a valuable tool for in vitro assays involving various cell models including those related to metabolic disorders Additionally it is utilized in animal studies to explore its potential effects on energy metabolism and lipid-related conditions Drug screening efforts employ it to evaluate new therapeutic candidates targeting metabolic pathways Experimental concentrations typically depend on the specific research design employed ensuring relevance and accuracy in findings