Learn More
AdipoGen Octadecanoylamino fluorescein
SDP
Catalog No. 502560348
Encompass_Preferred
Chemical. CAS 110698-53-8. Formula C38H47NO6. Molecular Weight 613.78. 5-Octadecanoylaminofluorescein is a lipophilic/amphiphilic fluorescent probe useful in monitoring membrane lipid behaviour and fluorescence resonance energy transfer applications in life cells. It binds at amphiphitic interfaces and may be used as probe for studies of interfacial micropolarity, micro fluidity and local pH. The label makes the cell membrane bright green. Spectral data lambdaex 497 nm lambdaem 520±10 nm in ethanol.
Catalog No. 50-256-0348 Supplier AdipoGen Supplier No. CDXO0013M100
Chemical. CAS 110698-53-8. Formula C38H47NO6. Molecular Weight 613.78. 5-Octadecanoylaminofluorescein is a lipophilic/amphiphilic fluorescent probe useful in monitoring membrane lipid behaviour and fluorescence resonance energy transfer applications in life cells. It binds at amphiphitic interfaces and may be used as probe for studies of interfacial micropolarity, micro fluidity and local pH. The label makes the cell membrane bright green. Spectral data lambdaex 497 nm lambdaem 520±10 nm in ethanol.