Learn More
Sigma Aldrich Octadecyl-Functionalized Silica Gel
Catalog No. 37763525G Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-763-525G 25 g
37-763-5100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 37-763-525G Supplier Sigma Aldrich Supplier No. 37763525G
Only null left
Add to Cart
Add to Cart
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Octadecyl-functionalized silica gel is generally used as a stationary phase for low- and medium-pressure reversed-phase liquid chromatography.
Specifications
MDL Number MFCD00166485
Quantity 25 g