Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Octadecyl Isocyanate 80.0+%, TCI America™
SDP

Catalog No. I0130250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0130250G 250 g
I013025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I0130250G Supplier TCI America Supplier No. I0130250G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 112-96-9
Molecular Formula C19H37NO
Molecular Weight (g/mol) 295.51
MDL Number MFCD00002041
InChI Key QWDQYHPOSSHSAW-UHFFFAOYSA-N
Synonym Isocyanic Acid Octadecyl Ester, Isocyanic Acid Stearyl Ester, Stearyl Isocyanate
PubChem CID 66988
IUPAC Name 1-isocyanatooctadecane
SMILES CCCCCCCCCCCCCCCCCCN=C=O

Specifications

Color Yellow
Boiling Point 220°C
Quantity 250 g
UN Number 2206
Formula Weight 295.51
Percent Purity ≥80.0% (GC)
Physical Form Liquid
Chemical Name or Material Octadecyl Isocyanate
EINECSNumber (2)-1692
RTECSNumber NR0140000
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.