Learn More
Medchemexpress LLC Octadecyl rhodamine B chloride | 65603-19-2 | MFCD00467892 | 98.7% | 731.49 g·mol⁻¹ | C46H67ClN2O3 | 100 MG
SDP
Catalog No. 5000282786
Encompass_Preferred
Octadecyl Rhodamine B chloride is an amphiphilic fluorescent probe for labeling and staining lipid membranes in membrane biology research. It binds to membranes and shows strong fluorescence in its monomeric state; in aqueous solution it can aggregate and self-quench. Common uses include membrane staining, fusion assays, membrane-binding studies, and quantitative fluorescence measurements when maintained below the critical aggregation concentration.
  • Amphiphilic fluorescent probe for membrane staining.
  • Monomer absorption/emission: 565 nm/585 nm in Triton X-100.
  • Forms aggregates in water that cause fluorescence quenching at high concentration.
  • Suitable for membrane-binding, fusion, and kinetic fluorescence assays.
  • Store sealed at 4°C and protect from moisture and light; in solvent store at -80°C for extended storage.
  • Available in multiple small pack sizes suitable for laboratory experiments.
Catalog No. 50-002-82786 Supplier Medchemexpress LLC Supplier No. HYW127703100MG
Octadecyl Rhodamine B chloride is an amphiphilic fluorescent probe for labeling and staining lipid membranes in membrane biology research. It binds to membranes and shows strong fluorescence in its monomeric state; in aqueous solution it can aggregate and self-quench. Common uses include membrane staining, fusion assays, membrane-binding studies, and quantitative fluorescence measurements when maintained below the critical aggregation concentration.
  • Amphiphilic fluorescent probe for membrane staining.
  • Monomer absorption/emission: 565 nm/585 nm in Triton X-100.
  • Forms aggregates in water that cause fluorescence quenching at high concentration.
  • Suitable for membrane-binding, fusion, and kinetic fluorescence assays.
  • Store sealed at 4°C and protect from moisture and light; in solvent store at -80°C for extended storage.
  • Available in multiple small pack sizes suitable for laboratory experiments.