Learn More
Medchemexpress LLC Octadecyl rhodamine B chloride | 65603-19-2 | MFCD00467892 | 98.7% | 731.49 g/mol | C46H67ClN2O3 | 50 MG
SDP
Catalog No. 5000283634
Encompass_Preferred
Octadecyl rhodamine B chloride is a lipophilic fluorescent dye used to stain lipid membranes and study membrane dynamics. The long alkyl chain anchors the dye to membranes while the rhodamine fluorophore provides strong fluorescence in appropriate solvents. It readily forms aggregates in aqueous media, which can lead to self-quenching; handle and dilute accordingly for fluorescence assays.
  • Lipophilic amphiphilic fluorescent probe suitable for membrane staining.
  • Strong fluorescence with excitation/emission in the ~556-585 nm range.
  • Soluble in DMSO; forms aggregates in aqueous solutions.
  • Useful for membrane fusion and lipid-membrane assays.
  • High purity appropriate for research applications.
Catalog No. 50-002-83634 Supplier Medchemexpress LLC Supplier No. HYW12770350MG
Octadecyl rhodamine B chloride is a lipophilic fluorescent dye used to stain lipid membranes and study membrane dynamics. The long alkyl chain anchors the dye to membranes while the rhodamine fluorophore provides strong fluorescence in appropriate solvents. It readily forms aggregates in aqueous media, which can lead to self-quenching; handle and dilute accordingly for fluorescence assays.
  • Lipophilic amphiphilic fluorescent probe suitable for membrane staining.
  • Strong fluorescence with excitation/emission in the ~556-585 nm range.
  • Soluble in DMSO; forms aggregates in aqueous solutions.
  • Useful for membrane fusion and lipid-membrane assays.
  • High purity appropriate for research applications.