Learn More
Medchemexpress LLC Octadecyl rhodamine B chloride | 65603-19-2 | MFCD00467892 | 99.6% | 731.49 g/mol | C46H67ClN2O3 | 25 MG
SDP
Catalog No. 5000283633
Encompass_Preferred
Octadecyl rhodamine B chloride is an amphiphilic fluorescent dye that incorporates into lipid membranes via its long octadecyl chain, enabling membrane labeling and quantitative analysis of membrane fusion and lipid-mixing events. Its fluorescence is concentration-dependent, allowing dequenching-based assays when dye is diluted in membranes.
  • Amphiphilic fluorescent probe for membrane labeling.
  • Partitions into lipid bilayers via an octadecyl alkyl chain.
  • Exhibits self-quenching at high local concentrations for dequenching assays.
  • Useful for lipid-mixing and membrane fusion studies.
  • Supplied as a high-purity solid suitable for experimental use.
Catalog No. 50-002-83633 Supplier Medchemexpress LLC Supplier No. HYW12770325MG
Octadecyl rhodamine B chloride is an amphiphilic fluorescent dye that incorporates into lipid membranes via its long octadecyl chain, enabling membrane labeling and quantitative analysis of membrane fusion and lipid-mixing events. Its fluorescence is concentration-dependent, allowing dequenching-based assays when dye is diluted in membranes.
  • Amphiphilic fluorescent probe for membrane labeling.
  • Partitions into lipid bilayers via an octadecyl alkyl chain.
  • Exhibits self-quenching at high local concentrations for dequenching assays.
  • Useful for lipid-mixing and membrane fusion studies.
  • Supplied as a high-purity solid suitable for experimental use.