Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Octadecyl Thioglycolate 50.0+%, TCI America™
SDP

Catalog No. T081125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T081125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T081125G Supplier TCI America Supplier No. T081125G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 10220-46-9
Molecular Formula C20H40O2S
Molecular Weight (g/mol) 344.60
MDL Number MFCD00022084
InChI Key JQTFPHLEQLLQOT-UHFFFAOYSA-N
Synonym Mercaptoacetic Acid Octadecyl Ester, Thioglycolic Acid Octadecyl Ester, Stearyl Thioglycolate, Thioglycolic Acid Stearyl Ester
PubChem CID 82462
IUPAC Name octadecyl 2-sulfanylacetate
SMILES CCCCCCCCCCCCCCCCCCOC(=O)CS

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 344.60
Percent Purity ≥50.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Octadecyl Thioglycolate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.