Learn More
Abcam 10-Octadecylacridine orange bromide, Fluorescent reagent, 20MG
Supplier:  Abcam AB14546920MG
Encompass_Preferred
MW 598.74 g/mol. Highly selective fluorescent reagent. Potential sensitive dye to monitor differences in cell potential as they occur during ion transport through the cell membrane.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-996-206