Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Octadecylamine

Catalog No. O14083KG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 g
3 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O14083KG 3 kg
O1408500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O14083KG Supplier Sigma Aldrich Supplier No. O14083KG
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Octadecylamine

Specifications

CAS 124-30-1
Melting Point 50°C to 52°C (lit.)
Boiling Point 232°C (32 mmHg)
Molecular Formula C18H39N
Linear Formula CH3(CH2)17 NH2
Quantity 3 kg
MDL Number MFCD00008159
Synonym 1-Aminooctadecane; Stearylamine
Molecular Weight (g/mol) 269.51
Percent Purity 90%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.