Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Octadecyltrimethoxysilane, 90%, Tech.

Catalog No. ac313600250
Change view
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC313600250 25 mL Glass bottle
AC313601000 100 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AC313600250 Supplier Thermo Scientific Chemicals Supplier No. 313600250
Only null left
Add to Cart
This item is not returnable. View return policy

CAS: 3069-42-9 | C21H46O3Si | 374.69 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3069-42-9
Molecular Formula C21H46O3Si
Molecular Weight (g/mol) 374.69
MDL Number MFCD00043060
InChI Key SLYCYWCVSGPDFR-UHFFFAOYSA-N
Synonym octadecyltrimethoxysilane, trimethoxy octadecyl silane, n-octadecyltrimethoxysilane, stearyltrimethoxysilane, silane, trimethoxyoctadecyl, octadecyl-trimethoxysilane, acmc-209hi7, ksc491g4l, trimethoxy octadecyl silane, technical grade
PubChem CID 76486
IUPAC Name trimethoxy(octadecyl)silane
SMILES CCCCCCCCCCCCCCCCCC[Si](OC)(OC)OC

Specifications

CAS Min % 88.0
CAS Max % 100.0
Melting Point 13°C to 17°C
Color Colorless to Yellow
Density 0.8800g/mL
Boiling Point 170°C (0.1 mmHg)
Flash Point 139°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. Total octadecyltrimethoxysilanes (GC)
Packaging Glass bottle
Refractive Index 1.4380 to 1.44
Linear Formula CH3(CH2)17Si(OCH3)3
Quantity 25 mL
Specific Gravity 0.88
Solubility Information Solubility in water: insoluble (reacts)
Formula Weight 374.69
Percent Purity 90%
Grade Technical
Physical Form Liquid
Chemical Name or Material Octadecyltrimethoxysilane
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 221-339-2
Recommended Storage Product may become turbid on storage

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.