Learn More
Cayman Chemical Octaethylene Glycol monododecyl ether, Purity: ≥95%, CAS: 3055-98-9, 1 g
SDP
Catalog No. 502980814
Encompass_Preferred
A nonionic surfactant used for solubilization of membrane-bound proteins
Catalog No. 50-298-0814 Supplier Cayman Chemical Supplier No. 209381 G
A nonionic surfactant used for solubilization of membrane-bound proteins