Learn More
Medchemexpress LLC Octaethylene glycol | 500mg
SDP
Catalog No. 5000209776
Encompass_Preferred
Octaethylene glycol is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs[1]
Catalog No. 50-002-09776 Supplier Medchemexpress LLC Supplier No. HYW050087500MG
Octaethylene glycol is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs[1]