Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Octaethylene glycol | 5117-19-1 | 100.0% | 370.44 | C16H34O9 | 1 G
SDP

Catalog No. 5000209775
Encompass_Preferred

Octaethylene glycol is a polyethylene glycol (PEG)-based linker used as a flexible, hydrophilic spacer in PROTAC synthesis and other chemical biology applications. It is commonly used to connect ligands in bifunctional molecules and enable targeted protein degradation via the ubiquitin-proteasome system.

  • Provides a flexible, hydrophilic spacer for bifunctional molecules.
  • Suitable for use as a PROTAC linker in chemical biology and medicinal chemistry.
  • High purity (99.97%) for research applications.
  • Molecular weight 370.44 and formula C16H34O9.
  • Appearance: solid below 22°C and liquid above 22°C; colorless to off-white.
  • Available in small-scale quantities including 1 g for laboratory use.

Catalog No. 50-002-09775 Supplier Medchemexpress LLC Supplier No. HYW0500871G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Octaethylene glycol is a polyethylene glycol (PEG)-based linker used as a flexible, hydrophilic spacer in PROTAC synthesis and other chemical biology applications. It is commonly used to connect ligands in bifunctional molecules and enable targeted protein degradation via the ubiquitin-proteasome system.

  • Provides a flexible, hydrophilic spacer for bifunctional molecules.
  • Suitable for use as a PROTAC linker in chemical biology and medicinal chemistry.
  • High purity (99.97%) for research applications.
  • Molecular weight 370.44 and formula C16H34O9.
  • Appearance: solid below 22°C and liquid above 22°C; colorless to off-white.
  • Available in small-scale quantities including 1 g for laboratory use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.