Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Octaethylene glycol | 5117-19-1 | MFCD00698694 | 100.0% | 370.44 g/mol | C16H34O9 | 5 G
SDP

Catalog No. 5000210615
Encompass_Preferred

Octaethylene glycol is a polyethylene glycol (PEG) linker composed of eight ethylene glycol units. It is a hydrophilic spacer used in chemical synthesis, commonly as a linker for proteolysis targeting chimeras (PROTACs) and other bioconjugation applications. The substance is a diol with terminal hydroxyl groups and is soluble in water and polar organic solvents.

  • Cas number 5117-19-1.
  • Molecular formula C16H34O9.
  • Molecular weight 370.44 g/mol.
  • High purity suitable for research use.
  • Functional groups: terminal hydroxyls for conjugation.
  • Uses: peg-based linker for PROTAC synthesis and bioconjugation.
  • Available in small lab-scale quantities (5 G pack).

Catalog No. 50-002-10615 Supplier Medchemexpress LLC Supplier No. HYW0500875G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Octaethylene glycol is a polyethylene glycol (PEG) linker composed of eight ethylene glycol units. It is a hydrophilic spacer used in chemical synthesis, commonly as a linker for proteolysis targeting chimeras (PROTACs) and other bioconjugation applications. The substance is a diol with terminal hydroxyl groups and is soluble in water and polar organic solvents.

  • Cas number 5117-19-1.
  • Molecular formula C16H34O9.
  • Molecular weight 370.44 g/mol.
  • High purity suitable for research use.
  • Functional groups: terminal hydroxyls for conjugation.
  • Uses: peg-based linker for PROTAC synthesis and bioconjugation.
  • Available in small lab-scale quantities (5 G pack).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.