Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Octaethylene glycol monododecyl ether (C12E8) | 3055-98-9 | 98.0% | 538.75 | 1 ML
SDP

Catalog No. 5000352108
Encompass_Preferred

Octaethylene glycol monododecyl ether, also known as C12E8, is a non-ionic detergent. It is utilized for the extraction of membrane proteins and possesses the ability to solubilize the viral membrane of intact influenza virus.

  • Functions as a non-ionic detergent.
  • Can be used for membrane protein extraction.
  • Capable of solubilizing the viral membrane of intact influenza virus.
  • Mechanism of solubilization involves the accumulation of molecules outside the membrane, leading to the formation of micelles and extraction of membrane constituents.
  • Available in liquid form with a density of 0.984-0.992 g/cm³.
  • Appears colorless to light yellow.
  • Has a purity of 98.0%.
  • Can be stored at -20°C for three years or 4°C for two years in pure form.
  • When in solvent, it can be stored at -80°C for six months or -20°C for one month.

Catalog No. 50-003-52108 Supplier Medchemexpress LLC Supplier No. HY13894110MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Octaethylene glycol monododecyl ether, also known as C12E8, is a non-ionic detergent. It is utilized for the extraction of membrane proteins and possesses the ability to solubilize the viral membrane of intact influenza virus.

  • Functions as a non-ionic detergent.
  • Can be used for membrane protein extraction.
  • Capable of solubilizing the viral membrane of intact influenza virus.
  • Mechanism of solubilization involves the accumulation of molecules outside the membrane, leading to the formation of micelles and extraction of membrane constituents.
  • Available in liquid form with a density of 0.984-0.992 g/cm³.
  • Appears colorless to light yellow.
  • Has a purity of 98.0%.
  • Can be stored at -20°C for three years or 4°C for two years in pure form.
  • When in solvent, it can be stored at -80°C for six months or -20°C for one month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.