Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Octafluoro-1,4-diiodobutane, 98%

Catalog No. AC426320050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC426320050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC426320050 Supplier Thermo Scientific Chemicals Supplier No. 426320050
  • $170.90
    $145.65 / Each of 1

    Save $25.25

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 375-50-8 | C4F8I2 | 453.84 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 375-50-8
Molecular Formula C4F8I2
Molecular Weight (g/mol) 453.84
MDL Number MFCD00042263
InChI Key JILAKKYYZPDQBE-UHFFFAOYSA-N
Synonym 1,4-diiodooctafluorobutane, octafluoro-1,4-diiodobutane, 1,4-diiodoperfluorobutane, perfluoro-1,4-diiodobutane, 1,4-diiodo octafluorobutane, butane, 1,1,2,2,3,3,4,4-octafluoro-1,4-diiodo, acmc-1aigi, i cf2 4i, 1,1,2,2,3,3,4,4-octafluoro-1,4-diiodo-butane
PubChem CID 78993
IUPAC Name 1,1,2,2,3,3,4,4-octafluoro-1,4-diiodobutane
SMILES FC(F)(I)C(F)(F)C(F)(F)C(F)(F)I

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -9.0°C
Color Colorless to Yellow or Pink
Density 2.4740g/mL
Boiling Point 150.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4280 to 1.4320 (20°C, 589nm)
Linear Formula CF2(I)(CF2)2CF2I
Quantity 5 g
Specific Gravity 2.474
Formula Weight 453.83
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Octafluoro-1, 4-diiodobutane
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause drowsiness or dizziness.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER
EINECSNumber 206-788-4

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.