Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Octafluorotoluene

Catalog No. O27065G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O27065G 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O27065G Supplier Sigma Aldrich Supplier No. O27065G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Physical properties (such as boiling point, refractive index and density) of 5-ethyl-1,3-dioxane-5-methanol have been reported.

Specifications

CAS 434-64-0
Density 1.666 g/mL (at 25°C (literature))
Boiling Point 104°C (lit.)
Molecular Formula C7F8
Refractive Index n20/D 1.368 (literature)
Linear Formula C6F5CF3
MDL Number MFCD00000375
Quantity 500 mL
Synonym Perfluorotoluene
Molecular Weight (g/mol) 236.06
Percent Purity 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.