Learn More
Cayman Chemical Octahydrocurcumin, Purity: ≥95%, CAS: 36062-07-4, 10 mg
SDP
Catalog No. 502949055
Encompass_Preferred
An active metabolite of curcumin with diverse biological activities; scavenges DPPH radicals in a cell-free assay (IC50 = 12.3 µM); inhibits LPS-induced increases in iNOS and COX-2 levels, production of NO, and nuclear translocation of NF-ᴋB in RAW 264.7 cells at 6.25 µM; active against various bacteria and fungi in disc assays; increases survival, reduces tumor weight, and induces tumor cell apoptosis in an H22 murine hepatocellular carcinoma model at 5, 10, and 20 mg/kg
Catalog No. 50-294-9055 Supplier Cayman Chemical Supplier No. 3368610 MG
An active metabolite of curcumin with diverse biological activities; scavenges DPPH radicals in a cell-free assay (IC50 = 12.3 µM); inhibits LPS-induced increases in iNOS and COX-2 levels, production of NO, and nuclear translocation of NF-ᴋB in RAW 264.7 cells at 6.25 µM; active against various bacteria and fungi in disc assays; increases survival, reduces tumor weight, and induces tumor cell apoptosis in an H22 murine hepatocellular carcinoma model at 5, 10, and 20 mg/kg