Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Octakis(dimethylsilyloxy)octasilsesquioxane, TCI America™
SDP

Catalog No. O03511G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O03511G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O03511G Supplier TCI America Supplier No. O03511G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 125756-69-6
Molecular Formula C16H48O20Si16
Molecular Weight (g/mol) 1009.9
MDL Number MFCD03701363
InChI Key NKAADCIRFJPCOM-UHFFFAOYSA-N
Synonym 1,3,5,7,9,11,13,15-Octakis(dimethylsilyloxy)pentacyclo[9.5.1.1(3,9).1(5,15).1(7,13)]octasiloxane
PubChem CID 11274685
SMILES C[Si](C)O[Si]12O[Si]3(O[Si]4(O[Si](O1)(O[Si]5(O[Si](O2)(O[Si](O3)(O[Si](O4)(O5)O[Si](C)C)O[Si](C)C)O[Si](C)C)O[Si](C)C)O[Si](C)C)O[Si](C)C)O[Si](C)C

Specifications

Melting Point 250°C
Color White
Quantity 1 g
Formula Weight 1017.96
Physical Form Crystalline Powder
Chemical Name or Material Octakis(dimethylsilyloxy)octasilsesquioxane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.