Learn More
Medchemexpress LLC Octamethylcyclotetra 25g
SDP
Catalog No. 5000206145
Encompass_Preferred
Octamethylcyclotetrasiloxane promotes the anchorage-independent growth of MCF-10A and MCF-10F cells Octamethylcyclotetrasiloxane induces DNA damage inhibits the expression of DNA-repairing protein BRCA1 under long-term and high-concentration exposure Octamethylcyclotetrasiloxane exhibits intrinsic estrogenic activity[1]
Catalog No. 50-002-06145 Supplier Medchemexpress LLC Supplier No. 25GHYW116007
Octamethylcyclotetrasiloxane promotes the anchorage-independent growth of MCF-10A and MCF-10F cells Octamethylcyclotetrasiloxane induces DNA damage inhibits the expression of DNA-repairing protein BRCA1 under long-term and high-concentration exposure Octamethylcyclotetrasiloxane exhibits intrinsic estrogenic activity[1]