Learn More
Medchemexpress LLC Octan-2-yl 2-cyanoac 50mg | 133978-15-1 | 209.28 g·mol⁻¹ | C12H19NO2 | 50 MG
SDP
Catalog No. 5000229310
Encompass_Preferred
Octan-2-yl 2-cyanoacrylate is a cyanoacrylate ester used as a fast-curing adhesive and tissue adhesive in research applications. Supplied for research use only, it polymerizes to form a flexible, durable film and is commonly used in adhesive testing, biomaterials research, and surgical adhesive studies.
  • Fast-curing cyanoacrylate adhesive for rapid bonding.
  • Forms a flexible, durable polymer film after curing.
  • Suitable for tissue adhesive research applications.
  • Available in small research quantities, improving handling for lab studies.
  • Characterized by datasheet, COA, and SDS for research validation.
  • Not for use in patients; intended for research use only.
Catalog No. 50-002-29310 Supplier Medchemexpress LLC Supplier No. HY11654450MG
Octan-2-yl 2-cyanoacrylate is a cyanoacrylate ester used as a fast-curing adhesive and tissue adhesive in research applications. Supplied for research use only, it polymerizes to form a flexible, durable film and is commonly used in adhesive testing, biomaterials research, and surgical adhesive studies.
  • Fast-curing cyanoacrylate adhesive for rapid bonding.
  • Forms a flexible, durable polymer film after curing.
  • Suitable for tissue adhesive research applications.
  • Available in small research quantities, improving handling for lab studies.
  • Characterized by datasheet, COA, and SDS for research validation.
  • Not for use in patients; intended for research use only.