Learn More
Medchemexpress LLC Octanal | 124-13-0 | 1 ML
SDP
Catalog No. 5000417332
Encompass_Preferred
Octanal is a fragrant aldehyde known for its antioxidant and antibacterial properties. It demonstrates antifungal effects against G. citri-aurantii by disrupting cell membrane integrity and causing leakage of cellular components. Additionally, it exhibits cytotoxicity towards HeLa cells.
  • Has antioxidant activity
  • Exhibits antibacterial activity
  • Shows antifungal effects against G. citri-aurantii
  • Increases extracellular conductivity and enhances membrane permeability in G. citri-aurantii mycelium
  • Decreases lipid content in G. citri-aurantii mycelium
  • Exhibits cytotoxicity towards HeLa cells
Catalog No. 50-004-17332 Supplier Medchemexpress LLC Supplier No. HYN801510MM/1ML
Octanal is a fragrant aldehyde known for its antioxidant and antibacterial properties. It demonstrates antifungal effects against G. citri-aurantii by disrupting cell membrane integrity and causing leakage of cellular components. Additionally, it exhibits cytotoxicity towards HeLa cells.
  • Has antioxidant activity
  • Exhibits antibacterial activity
  • Shows antifungal effects against G. citri-aurantii
  • Increases extracellular conductivity and enhances membrane permeability in G. citri-aurantii mycelium
  • Decreases lipid content in G. citri-aurantii mycelium
  • Exhibits cytotoxicity towards HeLa cells