Learn More
Medchemexpress LLC Octanal | 124-13-0 | 10 G
SDP
Catalog No. 5000419837
Encompass_Preferred
Octanal is a fragrant aldehyde with antioxidant and antibacterial activity. It exhibits antifungal effects against *G. citri-aurantii* by disrupting cell membrane integrity and causing cellular component leakage. It also shows cytotoxicity towards HeLa cells with an IC50 value of 3.5 μg/mL.
  • Antioxidant activity
  • Antibacterial activity
  • Antifungal effect against *G. citri-aurantii*
  • Cytotoxicity towards HeLa cells
Catalog No. 50-004-19837 Supplier Medchemexpress LLC Supplier No. HYN801510G
Octanal is a fragrant aldehyde with antioxidant and antibacterial activity. It exhibits antifungal effects against *G. citri-aurantii* by disrupting cell membrane integrity and causing cellular component leakage. It also shows cytotoxicity towards HeLa cells with an IC50 value of 3.5 μg/mL.
  • Antioxidant activity
  • Antibacterial activity
  • Antifungal effect against *G. citri-aurantii*
  • Cytotoxicity towards HeLa cells