Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Octanal | 124-13-0 | 25 G
SDP

Catalog No. 5000420738
Encompass_Preferred

Octanal is a fragrant aldehyde with antioxidant and antibacterial activity. Its antifungal effect against G. citri-aurantii may be related to the disruption of cell membrane integrity and the leakage of cellular components. Additionally, it exhibits cytotoxicity towards HeLa cells with an IC50 value of 3.5 μg/mL. This product is for research use only.

  • High purity (99.57%)
  • Molecular formula: C8H16O
  • Molecular weight: 128.22
  • Appears as a colorless to light yellow liquid
  • Target: Human endogenous metabolite
  • Demonstrates antifungal activity
  • Exhibits cytotoxicity towards HeLa cells

Catalog No. 50-004-20738 Supplier Medchemexpress LLC Supplier No. 25GHYN8015
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Octanal is a fragrant aldehyde with antioxidant and antibacterial activity. Its antifungal effect against G. citri-aurantii may be related to the disruption of cell membrane integrity and the leakage of cellular components. Additionally, it exhibits cytotoxicity towards HeLa cells with an IC50 value of 3.5 μg/mL. This product is for research use only.

  • High purity (99.57%)
  • Molecular formula: C8H16O
  • Molecular weight: 128.22
  • Appears as a colorless to light yellow liquid
  • Target: Human endogenous metabolite
  • Demonstrates antifungal activity
  • Exhibits cytotoxicity towards HeLa cells

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.