Learn More
Medchemexpress LLC Octanal-d16 | 1219794-66-7 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN8015S1MG
Encompass_Preferred
Octanal-d16 is the deuterium labeled Octanal. Octanal is a fragrant aldehyde with antioxidant and antibacterial activity. Its antifungal effect against *G. citri-aurantii* may be related to the disruption of cell membrane integrity and the leakage of cellular components. Additionally, Octanal exhibits cytotoxicity towards HeLa cells, with an IC50 value of 3.5 μg/mL.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-17261