Learn More
Medchemexpress LLC Octanal-d16 | 1219794-66-7 | 5 MG
SDP
Catalog No. 5000418158
Encompass_Preferred
Octanal-d16 is a deuterium-labeled octanal, a fragrant aldehyde known for its antioxidant and antibacterial properties. It has shown antifungal effects against *G. citri-aurantii* by disrupting cell membrane integrity and causing cellular component leakage. Furthermore, Octanal exhibits cytotoxicity towards HeLa cells, with an IC50 value of 3.5 μg/mL.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes have the potential to affect the pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-18158 Supplier Medchemexpress LLC Supplier No. HYN8015S5MG
Octanal-d16 is a deuterium-labeled octanal, a fragrant aldehyde known for its antioxidant and antibacterial properties. It has shown antifungal effects against *G. citri-aurantii* by disrupting cell membrane integrity and causing cellular component leakage. Furthermore, Octanal exhibits cytotoxicity towards HeLa cells, with an IC50 value of 3.5 μg/mL.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes have the potential to affect the pharmacokinetic and metabolic profiles of drugs