Learn More
Medchemexpress LLC Octanedioic-2,2,3,3,4,4,5,5,6,6,7,7-d12 acid | 169397-99-3 | 98.6% | 186.27 g/mol | C8H2D12O4 | 10 MG
SDP
Catalog No. 5000302624
Encompass_Preferred
Suberic acid-d12 is the deuterium-labeled isotopologue of suberic (octanedioic) acid used as an internal standard or tracer in analytical and biochemical applications. Supplied in small, pre-weighed quantities for laboratory use, the compound is characterized by high isotopic enrichment and manufacturer COA-verified purity.
  • Deuterium-labeled internal standard for mass spectrometry and tracer studies.
  • High isotopic enrichment (d12) for clear mass shift from native compound.
  • Manufacturer-verified purity of 98.6% suitable for analytical workflows.
  • Available in small, pre-weighed vials for direct use in assays.
  • Useful for quantitative analysis and metabolic tracing experiments.
Catalog No. 50-003-02624 Supplier Medchemexpress LLC Supplier No. HYW015300S110MG
Suberic acid-d12 is the deuterium-labeled isotopologue of suberic (octanedioic) acid used as an internal standard or tracer in analytical and biochemical applications. Supplied in small, pre-weighed quantities for laboratory use, the compound is characterized by high isotopic enrichment and manufacturer COA-verified purity.
  • Deuterium-labeled internal standard for mass spectrometry and tracer studies.
  • High isotopic enrichment (d12) for clear mass shift from native compound.
  • Manufacturer-verified purity of 98.6% suitable for analytical workflows.
  • Available in small, pre-weighed vials for direct use in assays.
  • Useful for quantitative analysis and metabolic tracing experiments.